2-Hydroxy-1-naphthaldehyde, tech.
2-Hydroxy-1-naphthaldehyde, tech.
2-Hydroxy-1-naphthaldehyde, tech.
Thermo Scientific Chemicals

2-Hydroxy-1-naphthaldehyde, tech.

CAS: 708-06-5 | C11H8O2 | 172.18 g/mol
Quantity:
100 g
500 g
Catalog number 121601000
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS708-06-5
IUPAC Name2-hydroxynaphthalene-1-carbaldehyde
Molecular FormulaC11H8O2
InChI KeyNTCCNERMXRIPTR-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Moist powder
Melting point>=77°C (on dry basis)
Appearance (Color)Yellow to brown
HPLC>=93 % (on dry basis)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only