1-Phenyl-1-cyclohexene, 95%, Thermo Scientific Chemicals
1-Phenyl-1-cyclohexene, 95%, Thermo Scientific Chemicals
1-Phenyl-1-cyclohexene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Phenyl-1-cyclohexene, 95%, Thermo Scientific Chemicals

Catalog NumberQuantityPackaging
13045025025 gGlass bottle
Catalog number 130450250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS771-98-2
IUPAC Name(cyclohex-1-en-1-yl)benzene
Molecular FormulaC12H14
InChI KeyWCMSFBRREKZZFL-UHFFFAOYSA-N
SMILESC1CCC(=CC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=94 %
Refractive index1.5680 to 1.5700 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only