trans-2-Phenylcyclopropane-1-carboxylic acid, 97%
trans-2-Phenylcyclopropane-1-carboxylic acid, 97%
trans-2-Phenylcyclopropane-1-carboxylic acid, 97%
Thermo Scientific Chemicals

trans-2-Phenylcyclopropane-1-carboxylic acid, 97%

CAS: 939-90-2 | C10H10O2 | 162.188 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 130460250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS939-90-2
IUPAC Name(1R,2R)-2-phenylcyclopropane-1-carboxylic acid
Molecular FormulaC10H10O2
InChI KeyAHDDRJBFJBDEPW-DTWKUNHWSA-N
SMILESOC(=O)[C@@H]1C[C@H]1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
GC>=96.0 %
Melting point88°C to 92°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only