Phenyl isocyanate, 99%
Phenyl isocyanate, 99%
Phenyl isocyanate, 99%
Thermo Scientific Chemicals

Phenyl isocyanate, 99%

CAS: 103-71-9 | C7H5NO | 119.12 g/mol
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 130692500
Price (JPY)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-71-9
IUPAC Nameisocyanatobenzene
Molecular FormulaC7H5NO
InChI KeyDGTNSSLYPYDJGL-UHFFFAOYSA-N
SMILESO=C=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Refractive index1.5350 to 1.5380 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only