3-Phenyl-5-isoxazolone +99%
3-Phenyl-5-isoxazolone +99%
3-Phenyl-5-isoxazolone +99%
Thermo Scientific Chemicals

3-Phenyl-5-isoxazolone +99%

CAS: 1076-59-1 | C9H7NO2 | 161.16 g/mol
Catalog NumberQuantityPackaging
1307000505 gGlass Bottle
Catalog number 130700050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1076-59-1
IUPAC Name3-phenyl-4,5-dihydro-1,2-oxazol-5-one
Molecular FormulaC9H7NO2
InChI KeyIHKNLPPRTQQACK-UHFFFAOYSA-N
SMILESO=C1CC(=NO1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline solid
HPLC>=99.0 %
Melting point150°C to 154°C
Appearance (Color)Light beige to pink
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only