Isopropyltriphenylphosphonium iodide, 98%
Isopropyltriphenylphosphonium iodide, 98%
Isopropyltriphenylphosphonium iodide, 98%
Thermo Scientific Chemicals

Isopropyltriphenylphosphonium iodide, 98%

CAS: 24470-78-8 | C21H22IP | 432.29 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 137081000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24470-78-8
IUPAC Nametriphenyl(propan-2-yl)phosphanium iodide
Molecular FormulaC21H22IP
InChI KeyHHBXWXJLQYJJBW-UHFFFAOYSA-M
SMILES[I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder or crystals
Appearance (Color)Light yellow to beige to light brown
Infrared spectrumConforms
Melting point195°C to 200°C
Titration Argentometric97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only