2-Thiophenemethanol, 99+%
2-Thiophenemethanol, 99+%
2-Thiophenemethanol, 99+%
Thermo Scientific Chemicals

2-Thiophenemethanol, 99+%

CAS: 636-72-6 | C5H6OS | 114.16 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 138861000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS636-72-6
IUPAC Name(thiophen-2-yl)methanol
Molecular FormulaC5H6OS
InChI KeyZPHGMBGIFODUMF-UHFFFAOYSA-N
SMILESOCC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5630 to 1.5660 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only