2-Chloro-6-nitrobenzaldehyde, 98%
2-Chloro-6-nitrobenzaldehyde, 98%
2-Chloro-6-nitrobenzaldehyde, 98%
Thermo Scientific Chemicals

2-Chloro-6-nitrobenzaldehyde, 98%

CAS: 6361-22-4 | C7H4ClNO3 | 185.56 g/mol
Quantity:
1 g
5 g
Catalog number 147750050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6361-22-4
IUPAC Name2-chloro-6-nitrobenzaldehyde
Molecular FormulaC7H4ClNO3
InChI KeyRZDOUWDCYULHJX-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(Cl)=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Infrared spectrumConforms
Melting point68°C to 73°C
GC>=97.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only