3-Methylpyridazine, 99%
3-Methylpyridazine, 99%
3-Methylpyridazine, 99%
Thermo Scientific Chemicals

3-Methylpyridazine, 99%

CAS: 1632-76-4 | C5H6N2 | 94.12 g/mol
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 148080050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1632-76-4
IUPAC Name3-methylpyridazine
Molecular FormulaC5H6N2
InChI KeyMXDRPNGTQDRKQM-UHFFFAOYSA-N
SMILESCC1=CC=CN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Refractive index1.5140 to 1.5160 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only