p-Tolunitrile, 98+%
p-Tolunitrile, 98+%
p-Tolunitrile, 98+%
Thermo Scientific Chemicals

p-Tolunitrile, 98+%

CAS: 104-85-8 | C8H7N | 117.15 g/mol
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 158020050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-85-8
IUPAC Name4-methylbenzonitrile
Molecular FormulaC8H7N
InChI KeyVCZNNAKNUVJVGX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Liquid or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only