2-Bromoethyl acetate, 97%
2-Bromoethyl acetate, 97%
2-Bromoethyl acetate, 97%
Thermo Scientific Chemicals

2-Bromoethyl acetate, 97%

CAS: 927-68-4 | C4H7BrO2 | 167.00 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 160091000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS927-68-4
IUPAC Name2-bromoethyl acetate
Molecular FormulaC4H7BrO2
InChI KeyRGHQKFQZGLKBCF-UHFFFAOYSA-N
SMILESCC(=O)OCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only