alpha-Bromo-m-tolunitrile, 95%
alpha-Bromo-m-tolunitrile, 95%
alpha-Bromo-m-tolunitrile, 95%
Thermo Scientific Chemicals

alpha-Bromo-m-tolunitrile, 95%

CAS: 28188-41-2 | C8H6BrN | 196.05 g/mol
Quantity:
10 g
50 g
Packaging:
View all
Glass bottle
Catalog number 163510100
Price (JPY)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS28188-41-2
IUPAC Name3-(bromomethyl)benzonitrile
Molecular FormulaC8H6BrN
InChI KeyCVKOOKPNCVYHNY-UHFFFAOYSA-N
SMILESBrCC1=CC=CC(=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=94 %
Melting point92°C to 97°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only