Product Image
Thermo Scientific Chemicals

Testosterone, Eur.Pharm., Thermo Scientific Chemicals

Testosterone, CAS # 58-22-0, is a steroid sex hormone from the androstane family and it is important to regulate and maintain several biological functions.
Catalog NumberQuantity
16441010010 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 164410100
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS58-22-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to almost white
Appearance (Color)to yellowish-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point153°C to 157°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Testosterone is a steroid sex hormone from the androstane family and is important for the maintenance of several biological functions

• This compound is an androgen that exerts its function by binding to and activating the androgen receptor

Applications

• Testosterone is irreversibly converted to dihydrotestosterone (DHT) in target tissues by the enzyme 5-∝ reductase

• Testosterone or DHT (its active metabolite) ligand-androgen receptor complexes act as a transcription factor, stimulating the expression of several responsive genes

• It antagonizes the androgen receptor to induce gene expression that causes the growth, development, and maintenance of several biological functions

• This compound is commonly used in vivo studies as an ester derivative due to its rapid metabolism by the liver

RUO – Research Use Only

Literature References

Tyagi, V.; Scordo, M.; Yoon, R.; Liporace, F.; Greene, L. Revisiting the role of testosterone: Are we missing something? Rev Urol. 2017, 19 (1), 16–24.