5-Chloro-2-pentanone ethylene ketal, 97%
5-Chloro-2-pentanone ethylene ketal, 97%
5-Chloro-2-pentanone ethylene ketal, 97%
5-Chloro-2-pentanone ethylene ketal, 97%
Thermo Scientific Chemicals

5-Chloro-2-pentanone ethylene ketal, 97%

CAS: 5978-08-5 | C7H13ClO2 | 164.63 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 166040250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5978-08-5
IUPAC Name2-(3-chloropropyl)-2-methyl-1,3-dioxolane
Molecular FormulaC7H13ClO2
InChI KeyOFERIRWCHSOJJT-UHFFFAOYSA-N
SMILESCC1(CCCCl)OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only