2-Methyl-2-nitropropane, 99%
2-Methyl-2-nitropropane, 99%
2-Methyl-2-nitropropane, 99%
Thermo Scientific Chemicals

2-Methyl-2-nitropropane, 99%

CAS: 594-70-7 | C4H9NO2 | 103.12 g/mol
Catalog number 182960250
Price (JPY)
-
Request bulk or custom format
Chemical Identifiers
CAS594-70-7
IUPAC Name2-methyl-2-nitropropane
Molecular FormulaC4H9NO2
InChI KeyAIMREYQYBFBEGQ-UHFFFAOYSA-N
SMILESCC(C)(C)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to white
Appearance (Form)Liquid or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only