L(+)-Isoleucinol, 97%
L(+)-Isoleucinol, 97%
L(+)-Isoleucinol, 97%
Thermo Scientific Chemicals

L(+)-Isoleucinol, 97%

CAS: 24629-25-2 | C6H16NO | 118.20 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 183940050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24629-25-2
IUPAC Name(2S,3S)-1-hydroxy-3-methylpentan-2-aminium
Molecular FormulaC6H16NO
InChI KeyVTQHAQXFSHDMHT-NTSWFWBYSA-O
SMILESCC[C@H](C)[C@H]([NH3+])CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Low melting mass and/or chunks or liquid
Melting point35°C to 40°C
Infrared spectrumConforms
Titration with HClO4>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only