Methanol-d4, for NMR, 100.0 atom % D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol-d4, for NMR, 100.0 atom % D, Thermo Scientific Chemicals

 
Catalog NumberQuantity
19048050050 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 190480500
Price (JPY)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS7784-34-1
IUPAC Nametrichloroarsane
Molecular FormulaAsCl3
InChI KeyOEYOHULQRFXULB-UHFFFAOYSA-N
SMILESCl[As](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Assay>=99.4 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only