Brij™ 30
Brij™ 30
Brij™ 30
Thermo Scientific Chemicals

Brij™ 30

CAS: 9002-92-0 | C14H30O2 | 230.39 g/mol
Quantity:
500 g
2.5 kg
Catalog number 216720025
Price (JPY)
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS9002-92-0
MDL NumberMFCD00043063
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Water=<1 %
Appearance (Form)Liquid or solid
Hydroxyl value145 to 165 mg KOH/g
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only