Oil, for heat transfer
Thermo Scientific Chemicals

Oil, for heat transfer

CAS: 64742-65- | C16H10N2Na2O7S2 | 452.363 g/mol
Quantity:
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 268770025
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS64742-65-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Appearance (Form)Oily liquid
Viscosity28.0 to 32.0 cSt (at 40°C) (kinematic)
Density(15°C) 0.84 to 0.88 kg/l
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only