Diallyl phthalate, 98%
Diallyl phthalate, 98%
Diallyl phthalate, 98%
Thermo Scientific Chemicals

Diallyl phthalate, 98%

CAS: 131-17-9 | C14H14O4 | 246.26 g/mol
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 276620025
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS131-17-9
IUPAC Name1,2-bis(prop-2-en-1-yl) benzene-1,2-dicarboxylate
Molecular FormulaC14H14O4
InChI KeyQUDWYFHPNIMBFC-UHFFFAOYSA-N
SMILESC=CCOC(=O)C1=CC=CC=C1C(=O)OCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.5180 to 1.5200 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only