Efficiently perform purification of high quality plasmid DNA or PCR products with the Thermo Scientific™ Nunc™ 96 Deep Well Filter Plates. Made of chemical resistant polypropylene material with superior mechanical strength, these plates conform to standard 96 well footprint compatible with high throughput sample processing.
Key Features:- Designed specifically for high throughput sample processing
- Shared wall configuration gives reduced plate height, wider wells for faster pocessing, higher yields and less clogging
- Suitable for vacuum, filtration, centrifugatioin or positive pressure techniques
- Polyethylene terapthalate (PET) frit in the fritted plates (278010, 278011) has 20μm pore size
- Glass Fiber plate (278010) has the PET frit plus two layers of glass fiber membrane for efficient binding of DNA
- Customized bar coding is available
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.