1-Hydroxybenzotriazole hydrate, 98%, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals

1-Hydroxybenzotriazole hydrate, 98%, Thermo Scientific Chemicals

 
Catalog NumberQuantityPackaging
30056025025 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 300560250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS123333-53-9
IUPAC Name1H-1,2,3-benzotriazol-1-ol
Molecular FormulaC6H5N3O
InChI KeyASOKPJOREAFHNY-UHFFFAOYSA-N
SMILESON1N=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH97.5 to 101.5 % (On dry substance)
Infrared spectrumConforms
HPLC>=97.5 %
Water11 to 26 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only