Dichlorophenylborane, 97%
Dichlorophenylborane, 97%
Dichlorophenylborane, 97%
Thermo Scientific Chemicals

Dichlorophenylborane, 97%

CAS: 873-51-8 | C6H5BCl2 | 158.82 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 318550250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS873-51-8
IUPAC Namedichloro(phenyl)borane
Molecular FormulaC6H5BCl2
InChI KeyNCQDQONETMHUMY-UHFFFAOYSA-N
SMILESClB(Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric96.0 to 104.0 % (ex Cl)
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only