Product Image
Thermo Scientific Chemicals

Diphenylphosphine, 95%, Thermo Scientific Chemicals

Catalog NumberQuantity
3209700505 g
32097025025 g
Catalog number 320970050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS829-85-6
IUPAC Namediphenylphosphane
Molecular FormulaC12H11P
InChI KeyGPAYUJZHTULNBE-UHFFFAOYSA-N
SMILESP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Infrared spectrumConforms
Refractive index1.6250 to 1.6300 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only