4-Methylumbelliferyl-alpha-D-galactopyranoside hydrate, 99%
4-Methylumbelliferyl-alpha-D-galactopyranoside hydrate, 99%
4-Methylumbelliferyl-alpha-D-galactopyranoside hydrate, 99%
Thermo Scientific Chemicals

4-Methylumbelliferyl-alpha-D-galactopyranoside hydrate, 99%

CAS: 38597-12-5 | C16H18O8 | 338.31 g/mol
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Catalog number 337160500
Price (JPY)
-
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS38597-12-5
IUPAC Name4-methyl-7-{[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC16H18O8
InChI KeyYUDPTGPSBJVHCN-HWSQZMMLNA-N
SMILESCC1=CC(=O)OC2=CC(O[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Water=<8 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only