ART™ Extended Length Tips
Thermo Scientific™
ART™ Extended Length Tips
Superior tips for any application, any pipette
Catalog NumberVolume (Metric)
3510PK
also known as 3510
10 μL
Catalog number 3510PK
also known as 3510
Price (JPY)
3,700
Pack of 1000
Volume (Metric):
10 μL
Superior tips for any application, any pipette
Thermo Scientific™ ART™ Extended Length Tips deliver proven ART quality, performance and reliability,
backed by the highest Thermo Scientific standards and worldwide support.
Every tip is certified to meet the industry's highest standards for molding precision and clarity, and are certified free of RNase, DNase, ATP, and Pyrogens. And each meets the exact specifications of its respective pipette brand manufacturer.
Specifications
DescriptionART 10 REACH, Non-Filtered Tip
FilteredNon-filtered
FormatBulk
Product LineART
SterilityNon-sterile
Volume (Metric)10 μL
Unit SizePack of 1000