2-Acetamido-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals
2-Acetamido-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals
2-Acetamido-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Acetamido-5-bromobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog NumberQuantity
35402025025 g
Catalog number 354020250
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS38985-79-4
IUPAC Name5-bromo-2-acetamidobenzoate
Molecular FormulaC9H7BrNO3
InChI KeyQVABAFHRLMDDLM-UHFFFAOYSA-M
SMILESCC(=O)NC1=CC=C(Br)C=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light brown
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
HPLC>=96.0 %
Melting point218°C to 223°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only