Product Image
Thermo Scientific Chemicals

Tri-tert-butylphosphine, 95%, Thermo Scientific Chemicals

Catalog NumberQuantity
3608900505 g
3608900101 g
Catalog number 360890050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS13716-12-6
IUPAC Nametri-tert-butylphosphane
Molecular FormulaC12H27P
InChI KeyBWHDROKFUHTORW-UHFFFAOYSA-N
SMILESCC(C)(C)P(C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
NMR>=94 %
Infrared spectrumConforms
Appearance (Color)Colorless
Appearance (Form)Low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only