Product Image
Thermo Scientific Chemicals

Carvacrol, Thermo Scientific Chemicals

Catalog NumberQuantity
36834025025 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 368340250
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS499-75-2
IUPAC Name2-methyl-5-(propan-2-yl)phenol
Molecular FormulaC10H14O
InChI KeyRECUKUPTGUEGMW-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear yellow to orange
Infrared spectrumConforms
Refractive index1.5220 to 1.5240 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only