Collodion, Thermo Scientific Chemicals
Collodion, Thermo Scientific Chemicals
Collodion, Thermo Scientific Chemicals
Collodion, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Collodion, Thermo Scientific Chemicals

Catalog NumberQuantityPackaging
405355001500 mLGlass bottle
Catalog number 405355001
Price (JPY)
-
Request A Quote
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS9004-70-0
IUPAC Name2,3-bis(nitrooxy)-6-[(nitrooxy)methyl]-5-{[3,4,5-tris(nitrooxy)-6-[(nitrooxy)methyl]oxan-2-yl]oxy}oxan-4-yl nitrate; 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC24H36N8O38
InChI KeyOWWGYSXLINVAES-UHFFFAOYNA-N
SMILESOCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O.[O-][N+](=O)OCC1OC(O[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1OC1OC(CO[N+]([O-])=O)C(O[N+]([O-])=O)C(O[N+]([O-])=O)C1O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
CompositionCastor oil: 5%
Infrared spectrumConforms
Density(25/4°C) 0.775 to 0.790 kg/l
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use