1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific Chemicals
1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific Chemicals
1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(2-Furyl)-1,3-butanedione, 96%, Thermo Scientific Chemicals

Catalog NumberQuantityPackaging
41083025025 gGlass bottle
Catalog number 410830250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25790-35-6
IUPAC Name1-(furan-2-yl)butane-1,3-dione
Molecular FormulaC8H8O3
InChI KeyGPYKJDYMMUIUFG-UHFFFAOYSA-N
SMILESCC(=O)CC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange-brown
Appearance (Form)Liquid after melting
Infrared spectrumConforms
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only