4-Fluorobenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™
4-Fluorobenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™
4-Fluorobenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™
Thermo Scientific Chemicals

4-Fluorobenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™

CAS: 1643-73-8 | C7H6ClFMg | 168.88 g/mol
Catalog NumberQuantity
42679050050 mL
Catalog number 426790500
Price (JPY)
-
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS1643-73-8
IUPAC Namechloro[(4-fluorophenyl)methyl]magnesium
Molecular FormulaC7H6ClFMg
InChI KeyPQDGQUPDDGUKLP-UHFFFAOYSA-M
SMILESFC1=CC=C(C[Mg]Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.15 to 0.35 M
Appearance (Form)Solution
Appearance (Color)Yellow to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only