2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98%
2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98%
2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98%
2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98%
Thermo Scientific Chemicals

2,2'-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline], 98%

CAS: 131833-93-7 | C17H30N2O2 | 294.44 g/mol
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 427520010
Price (JPY)
-
Request A Quote
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS131833-93-7
IUPAC Name4-tert-butyl-2-[2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Molecular FormulaC17H30N2O2
InChI KeyDPMGLJUMNRDNMX-UHFFFAOYNA-N
SMILESCC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Specific optical rotation-118° to -122° (20°C, 589 nm) (c=5, in CHCl3)
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only