4-Ethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
4-Ethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
4-Ethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

4-Ethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 22873-28-5 | C8H9BrMg | 209.37 g/mol
Catalog NumberQuantityPackaging
43189050050 mLAcroSeal™ Glass Bottle
Catalog number 431890500
Price (JPY)
-
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS22873-28-5
IUPAC Namemagnesium(2+) 4-ethylbenzen-1-ide bromide
Molecular FormulaC8H9BrMg
InChI KeyQSGSHZBZGXYVOQ-UHFFFAOYSA-M
SMILES[Mg++].[Br-].CCC1=CC=[C-]C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration Grignard0.40 to 0.60 M
AppearanceMay develop some turbidity or precipitate
Appearance (Color)CLear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only