Tetrabutylammonium fluoride, 75% solution in water
Tetrabutylammonium fluoride, 75% solution in water
Tetrabutylammonium fluoride, 75% solution in water
Tetrabutylammonium fluoride, 75% solution in water
Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 75% solution in water

CAS: 429-41-4 | C16H36FN | 261.47 g/mol
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 451801000
Price (JPY)
-
Request A Quote
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS429-41-4
IUPAC Nametetrabutylazanium fluoride
Molecular FormulaC16H36FN
InChI KeyFPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES[F-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Titration with HClO470 to 80 %
Appearance (Form)Viscous liquid
Water20 to 30 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only