Product Image
Thermo Scientific Chemicals

Moxalactam sodium salt

CAS: 64953-12-4 | C20H18N6Na2O9S | 564.44 g/mol
Catalog NumberQuantity
4604100101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460410010
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS64953-12-4
IUPAC Namedisodium 7-[2-carboxylato-2-(4-hydroxyphenyl)acetamido]-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular FormulaC20H18N6Na2O9S
InChI KeyGRIXGZQULWMCLU-UHFFFAOYNA-L
SMILES[Na+].[Na+].COC1(NC(=O)C(C([O-])=O)C2=CC=C(O)C=C2)C2OCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Potency>=830 µg/mg (on anhydrous basis)
Water=<5.0 % (K.F.)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only