Chemical Identifiers
CAS90357-06-5
IUPAC NameN-[4-cyano-3-(trifluoromethyl)phenyl]-3-(4-fluorobenzenesulfonyl)-2-hydroxy-2-methylpropanamide
Molecular FormulaC18H14F4N2O4S
InChI KeyLKJPYSCBVHEWIU-UHFFFAOYNA-N
SMILESCC(O)(CS(=O)(=O)C1=CC=C(F)C=C1)C(=O)NC1=CC=C(C#N)C(=C1)C(F)(F)F
View more