4-Cyanoimidazole, 95%
4-Cyanoimidazole, 95%
4-Cyanoimidazole, 95%
Thermo Scientific Chemicals

4-Cyanoimidazole, 95%

CAS: 57090-88-7 | C4H3N3 | 93.09 g/mol
Quantity:
1 g
5 g
Catalog number 461360050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS57090-88-7
IUPAC Name1H-imidazole-5-carbonitrile
Molecular FormulaC4H3N3
InChI KeyNWVGXXPWOYZODV-UHFFFAOYSA-N
SMILESN#CC1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point141°C to 148°C
HPLC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only