Product Image
Thermo Scientific Chemicals

Sitafloxacin sesquihydrate, Thermo Scientific Chemicals

Catalog NumberQuantity
464372500250 mg
4643700101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464372500
Price (JPY)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS163253-35-8
IUPAC Namebis(7-{7-amino-5-azaspiro[2.4]heptan-5-yl}-8-chloro-6-fluoro-1-(2-fluorocyclopropyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid) trihydrate
Molecular FormulaC38H42Cl2F4N6O9
InChI KeyMPORYQCGWFQFLA-UHFFFAOYNA-N
SMILESO.O.O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O.NC1CN(CC11CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3F)C2=C1Cl)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Loss on drying5.5 % to 8.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only