Product Image
Thermo Scientific Chemicals

Silver proteinate, mild, ∼20% silver

Quantity:
5 g
25 g
Catalog number 466400050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1301199-90-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown or black
Identification APasses test
Identification BPasses test
Silver (Ag)19 to 23 %
Appearance of solution(20 mg/ml, H2O) Opaque, brown solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only