Search
| Catalog Number | Color |
|---|---|
| 8870PK | Fluorescent Pink |
| 8860PK also known as 8860 | Fluorescent Orange |
| 8850PK also known as 8850 | Fluorescent Green |
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.