Acetic Anhydride, Certified AR for Analysis, Fisher Chemical™
Acetic Anhydride, Certified AR for Analysis, Fisher Chemical™
Acetic Anhydride, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Acetic Anhydride, Certified AR for Analysis, Fisher Chemical™

Quantity:
500 mL
2.5 L
Catalog number A-0480-PB08
also known as A/0480/PB08
Price (JPY)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
CAS108-24-7
ColorColorless
Formula Weight102.09g/mol
MDL Number8705
Melting Point-73.1°C
View more
Acetic Anhydride, Certified AR for Analysis