Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
Fisher Chemical™

Acetone, Certified AR for analysis, meets analytical specification of Ph.Eur, Fisher Chemical™

C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, CHEBI:15347, Colorless, 58.08g/mol, CSCPPACGZOOCGX-UHFFFAOYSA-N, propan-2-one, -95 deg.C, 8765, Amber glass bottle, Liquid, 180, CC(-O)C, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C | CAS: 67-64-1 | C3H6O | 58.08 g/mol
Quantity:
2.5 L
5 L
Packaging:
View all
Amber Glass Bottle
HDPE Plastic Bottle
Catalog number A-0600-17
also known as A/0600/17
Price (JPY)
-
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS67-64-1
Specifications
Boiling Point56°C
CAS67-64-1
ChEBICHEBI:15347
ColorColorless
Formula Weight58.08g/mol
View more
C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, HDPE plastic bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C