Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™
Fisher Chemical™

Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™

Catalog NumberQuantity
A-5360-53
also known as A/5360/53
500 g
Catalog number A-5360-53
also known as A/5360/53
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Specifications
CAS7783-28-0
ColorWhite
Formula Weight132.06g/mol
IUPAC Namephosphoric acid diamine
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
View more
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis