You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Chemicals
▸
Salts and Inorganics
▸
Inorganic Salts
▸
Ammonium Salts
▸
Ammonium Molybdate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical™
Ammonium Molybdate Tetrahydrate, Extra Pure, SLR, Fisher Chemical™
Technical Support
Customer Service
Catalog Number
Quantity
A-5720-48
also known as A/5720/48
100 g
Try the chem dex
TM
web app
Catalog number
A-5720-48
also known as A/5720/48
Price (JPY)
-
Contact Us ›
Quantity:
100 g
Request bulk or custom format
Specifications
CAS
12054-85-2
Color
White
Formula Weight
1235.86g/mol
IUPAC Name
ammonium hydrate molybdenum oxidandiide
InChI Key
XCOMAVCHADVANQ-UHFFFAOYSA-O
View more
Ammonium Molybdate Tetrahydrate, Extra Pure, SLR