Ammonium Nitrate, Certified AR for Analysis, Fisher Chemical™
Ammonium Nitrate, Certified AR for Analysis, Fisher Chemical™
Ammonium Nitrate, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Ammonium Nitrate, Certified AR for Analysis, Fisher Chemical™

H4N2O3, CAS Number-6484-52-2, 500g, 210 deg.C, White, 80.04g/mol, 169 deg.C, 11425, Plastic powder jar, 4.5 to 6.0, Solid
Quantity:
500 g
1 kg
Catalog number A-5920-60
also known as A/5920/60
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
Boiling Point210°C
CAS6484-52-2
ColorWhite
Formula Weight80.04g/mol
IUPAC Nameammonium nitrate
View more
H4N2O3, CAS Number-6484-52-2, 500g, 210 deg.C, White, 80.04g/mol, 169 deg.C, 11425, Plastic powder jar, 4.5 to 6.0, Solid