3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%
Thermo Scientific Chemicals

3-Chloro-5,5-dimethyl-2-cyclohexen-1-one, 96%

CAS: 17530-69-7 | C8H11ClO | 158.63 g/mol
Catalog NumberQuantity
A10728.22
also known as A10728-22
100 g
Catalog number A10728.22
also known as A10728-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS17530-69-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.4885-1.4945 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only