Cholesterol, 95%, Thermo Scientific Chemicals
Cholesterol, 95%, Thermo Scientific Chemicals
Cholesterol, 95%, Thermo Scientific Chemicals
Cholesterol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cholesterol, 95%, Thermo Scientific Chemicals

A laboratory purified preparation of the animal sterol, Cholesterol, CAS # 57-88-5, is available in powder form for experimental use.
Quantity:
50 g
250 g
1000 g
Catalog number A11470.18
also known as A11470-18
Price (JPY)
-
Request A Quote
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS57-88-5
IUPAC Name(1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Molecular FormulaC27H46O
InChI KeyHVYWMOMLDIMFJA-DPAQBDIFSA-N
SMILESCC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥94.0% (non-U.S. specification)
Assay from Suppliers CofA≥94.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Cholesterol is used in the preparation of liposome. It is used to control the functions of transporters and coordinating cell proteins present in plasma membrane. It acts as a part of cell membranes. It benefits in rapid diagnostics. It is a component of all biological membranes and inhibits basal channel activity in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
  • Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
  • A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
  • It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems

Applications

  • Suitable for in vitro and in vivo applications
  • Stabilized in powder form for laboratory use only
RUO – Research Use Only

General References:

  1. Nierzwicki, L.; Czub, J. Specific Binding of Cholesetrol to the Amyloid Precursor Protein: Structure of the Complex and Driving Forces Characterized in Molecular Detail. J. Phys. Chem. Lett. 2015, 6 (5), 784-790.
  2. Lee, S. S.; Li, J.; Tai, J. N.; Ratliff, T. L.; Park, K.; Cheng, J. Avasimibe Encapsulated in Human Serum Albumin Blocks Cholesterol Esterification for Selective Cancer Treatment. ACS Nano. 2015, 9 (3), 2420-2432.