1,1-Bis(methylthio)-2-nitroethylene, 99%
1,1-Bis(methylthio)-2-nitroethylene, 99%
1,1-Bis(methylthio)-2-nitroethylene, 99%
Thermo Scientific Chemicals

1,1-Bis(methylthio)-2-nitroethylene, 99%

CAS: 13623-94-4 | C4H7NO2S2 | 165.225 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A11789.14
also known as A11789-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13623-94-4
IUPAC Name1,1-bis(methylsulfanyl)-2-nitroethene
Molecular FormulaC4H7NO2S2
InChI KeyNXGHEDHQXXXTTP-UHFFFAOYSA-N
SMILESCSC(SC)=C[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (GC)>98.5%
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only