2-Chloro-1-fluoro-4-iodobenzene, 98%
2-Chloro-1-fluoro-4-iodobenzene, 98%
2-Chloro-1-fluoro-4-iodobenzene, 98%
Thermo Scientific Chemicals

2-Chloro-1-fluoro-4-iodobenzene, 98%

CAS: 156150-67-3 | C6H3ClFI | 256.443 g/mol
Quantity:
25 g
100 g
Catalog number A11808.14
also known as A11808-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS156150-67-3
IUPAC Name2-chloro-1-fluoro-4-iodobenzene
Molecular FormulaC6H3ClFI
InChI KeyOMASDGWBVAVFQZ-UHFFFAOYSA-N
SMILESFC1=C(Cl)C=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6020-1.6060 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
It employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Victoria E. Laing. et.al. Fused thiophene derivatives as MEK inhibitors. Bioorganic & Medicinal Chemistry Letters.,2012, 22(1), 472-475.